trans-1-Boc-4-fluoro-3-hydroxypiperidine

Supplier: Matrix Scientific

Synonyms: trans-tert-Butyl 4-fluoro-3-hydroxypiperidine-1-carboxylate

113392-1G 113392-5G
102892-048EA 867.69 USD
102892-048 102892-050
trans-1-Boc-4-fluoro-3-hydroxypiperidine
trans-1-Boc-4-fluoro-3-hydroxypiperidine

Hazard: Irritant

Formula: C₁₀H₁₈FNO₃
MW: 219.26 g/mol
MDL Number: MFCD18425630
CAS Number: 955028-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR