Azetidine-1,3,3-tricarboxylic acid-1-tert-butyl ester-3-ethyl ester

Supplier: Matrix Scientific

113390-1G 113390-5G
102892-040EA 844.62 USD
102892-040 102892-042
Azetidine-1,3,3-tricarboxylic acid-1-tert-butyl ester-3-ethyl ester
Azetidine-1,3,3-tricarboxylic acid-1-tert-butyl ester-3-ethyl ester

Hazard: Irritant

Formula: C₁₂H₁₉NO₆
MW: 273.29 g/mol
MDL Number: MFCD18374994
CAS Number: 1011479-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR