Methyl 4-aminooxane-4-carboxylate hydrochloride

Supplier: Matrix Scientific

Synonyms: Methyl-4-aminotetrahydro-2H-pyran-4-carboxylate hydrochloride

113389-1G 113389-5G
102892-036EA 581.54 USD
102892-036 102892-038
Methyl 4-aminooxane-4-carboxylate hydrochloride
Methyl 4-aminooxane-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₇H₁₃NO₃·ClH
MW: 195.644 g/mol
MDL Number: MFCD11226845
CAS Number: 199330-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR