Ethyl (tetrahydropyran-4-ylidene)acetate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(dihydro-2H-pyran-4(3H)-ylidene)acetate

113388-1G 113388-5G
102892-032EA 233.85 USD
102892-032 102892-034
Ethyl (tetrahydropyran-4-ylidene)acetate
Ethyl (tetrahydropyran-4-ylidene)acetate

Hazard: Irritant

Formula: C₉H₁₄O₃
MW: 170.2057 g/mol
MDL Number: MFCD09475810
CAS Number: 130312-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR