7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)

Supplier: Matrix Scientific

Synonyms: 7-Oxa-2-azaspiro[3.5]nonane oxalate(2:1), 7-oxa-2-azaspiro[3.5]nonane hemioxalate

113387-1G 113387-5G
102892-028EA 407.69 USD
102892-028 102892-030
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)

Hazard: Irritant

Formula: C₁₆H₂₈N₂O₆
MW: 344.41 g/mol
MDL Number: MFCD22422285
CAS Number: 1523571-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR