3-(Chloromethyl)-5-fluoropyridine hydrochloride

Supplier: Matrix Scientific

113385-1G 113385-5G
102892-022EA 1018.46 USD
102892-022 102892-024
3-(Chloromethyl)-5-fluoropyridine hydrochloride
3-(Chloromethyl)-5-fluoropyridine hydrochloride

Hazard: Irritant

Formula: C₆H₆Cl₂FN
MDL Number: MFCD19687170
CAS Number: 1222556-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR