5-Bromo-2,3-pyridinedicarboxylic anhydride
Supplier: Matrix Scientific
Synonyms:
3-Bromofuro[3,4-b]pyridine-5,7-dione
113384-1G
113384-5G
102892-018EA
436.92
USD
102892-018
102892-020
5-Bromo-2,3-pyridinedicarboxylic anhydride
5-Bromo-2,3-pyridinedicarboxylic anhydride
Hazard: Irritant
Formula:
C₇H₂BrNO₃ MW: 228.01 g/mol |
MDL Number:
MFCD16250351 CAS Number: 98278-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...