8-Boc-2,8-diazaspiro[4.5]decane oxalate

Supplier: Matrix Scientific

Synonyms: 2,8-Diazaspiro[4.5]decane oxalate, N8-BOC protected

113377-1G 113377-5G
102892-002EA 727.69 USD
102892-002 102892-004
8-Boc-2,8-diazaspiro[4.5]decane oxalate
8-Boc-2,8-diazaspiro[4.5]decane oxalate

Hazard: Irritant

Formula: C₁₅H₂₆N₂O₆
MW: 330.38 g/mol
MDL Number: MFCD20527269
CAS Number: 1408074-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR