3-Bromo-1-tosyl-1H-pyrazolo[3,4-b]pyridine

Supplier: Matrix Scientific

113365-1G 113365-5G
102891-988EA 546.15 USD
102891-988 102891-990
3-Bromo-1-tosyl-1H-pyrazolo[3,4-b]pyridine
3-Bromo-1-tosyl-1H-pyrazolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₁₃H₁₀BrN₃O₂S
MDL Number: MFCD22628107
CAS Number: 889451-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR