N-Ethyloxetan-3-amine
Supplier: Matrix Scientific
113326-1G
113326-5G
102891-934EA
960
USD
102891-934
102891-936
N-Ethyloxetan-3-amine
N-Ethyloxetan-3-amine
Hazard: Irritant
Formula:
C₅H₁₁NO |
MDL Number:
MFCD18849120 CAS Number: 1341989-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...