4-Methyl-7H-pyrrolo[2,3-d]pyrimidine

Supplier: Matrix Scientific

113320-1G 113320-5G
102891-922EA 727.69 USD
102891-922 102891-924
4-Methyl-7H-pyrrolo[2,3-d]pyrimidine
4-Methyl-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₇H₇N₃
MDL Number: MFCD13619883
CAS Number: 945950-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR