3,3-Dimethylindoline

Supplier: Matrix Scientific

113317-1G 113317-5G
102897-744EA 436.92 USD
102897-744 102897-746
3,3-Dimethylindoline
3,3-Dimethylindoline

Hazard: Irritant

Formula: C₁₀H₁₃N
Boiling Pt: 60 °C (2 Torr)
Melting Pt: 31…33 °C
MDL Number: MFCD11847210
CAS Number: 1914-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR