Ethyl-1-boc-3-azetidineacetate

Supplier: Matrix Scientific

Synonyms: 3-(2-Ethoxy-2-oxoethyl)azetidine-1-carboxylic acid tert-Butyl ester, Ethyl 1-(tert-Butoxycarbonyl)-3-azetidineacetate, 1-(tert-Butoxycarbonyl)-3-(2-ethoxy-2-oxoethyl)azetidine, 1-Boc-3-(2-ethoxy-2-oxoethyl)azetidine, tert-Butyl 3-(2-Ethoxy-2-oxoethyl)azetidine-1-carboxylate, Ethyl 1-Boc-3-azetidineacetate, 1-(tert-Butoxycarbonyl)-3-azetidineacetic acid ethyl ester

113304-1G 113304-5G
102891-884EA 436.92 USD
102891-884 102891-886
Ethyl-1-boc-3-azetidineacetate
Ethyl-1-boc-3-azetidineacetate

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
MDL Number: MFCD11616049
CAS Number: 158602-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR