(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (2R)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected, (R)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

113302-1G 113302-5G
102891-876EA 436.92 USD
102891-876 102891-878
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid
(R)-1-Boc-4-Oxopiperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₅
MW: 243.26 g/mol
MDL Number: MFCD09953681
CAS Number: 1212176-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR