6-Boc-2-oxa-6-azaspiro[3.3]heptane

Supplier: Matrix Scientific

113299-1G 113299-5G
102891-872EA 436.92 USD
102891-872 102891-874
6-Boc-2-oxa-6-azaspiro[3.3]heptane
6-Boc-2-oxa-6-azaspiro[3.3]heptane

Hazard: Irritant

Formula: C₁₀H₁₇NO₃
MDL Number: MFCD12545889
CAS Number: 1263285-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR