1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde

Supplier: Matrix Scientific

113293-1G 113293-5G
102891-866EA 436.92 USD
102891-866 102891-868
1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde
1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₅H₁₁NO₃S
MW: 285.32 g/mol
MDL Number: MFCD03086091
CAS Number: 80360-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR