(R)-1-Boc-3-Methylaminopieridine

Supplier: Matrix Scientific

113277-1G 113277-5G
102891-814EA 872.31 USD
102891-814 102891-816
(R)-1-Boc-3-Methylaminopieridine
(R)-1-Boc-3-Methylaminopieridine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD09752963
CAS Number: 203941-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR