1-Boc-2-methyl-L-proline

Supplier: Matrix Scientific

Synonyms: (S)-1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid, Boc-α-Me-Pro-OH

113274-1G 113274-5G
102891-806EA 581.54 USD
102891-806 102891-808
1-Boc-2-methyl-L-proline
1-Boc-2-methyl-L-proline

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 134…138 °C
MDL Number: MFCD08706361
CAS Number: 103336-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR