2-Methyl-2H-indazole-3-methanol

Supplier: Matrix Scientific

Synonyms: (2-Methyl-2H-indazol-3-yl)methanol

113262-1G 113262-5G
102891-782EA 436.92 USD
102891-782 102891-784
2-Methyl-2H-indazole-3-methanol
2-Methyl-2H-indazole-3-methanol

Hazard: Irritant

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
MDL Number: MFCD08060471
CAS Number: 58536-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR