4,4-Difluoro-L-proline methyl ester trifluoroacetate

Supplier: Matrix Scientific

113261-1G 113261-5G
102891-754EA 727.69 USD
102891-754 102891-780
4,4-Difluoro-L-proline methyl ester trifluoroacetate
4,4-Difluoro-L-proline methyl ester trifluoroacetate

Hazard: Irritant

Formula: C₈H₁₀F₅NO₄
MDL Number: MFCD22415243
CAS Number: 915230-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR