1-Methylcyclobutanecarboxylic acid

Supplier: Matrix Scientific

113255-1G 113255-5G
102897-794EA 436.92 USD
102897-794 102897-796
1-Methylcyclobutanecarboxylic acid
1-Methylcyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD15527494
CAS Number: 32936-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR