3-Fluoro-4-piperidinone hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-piperidone Hydrochloride

113237-1G 113237-5G
102891-722EA 436.92 USD
102891-722 102891-724
3-Fluoro-4-piperidinone hydrochloride
3-Fluoro-4-piperidinone hydrochloride

Hazard: Irritant

Formula: C₅H₉ClFNO
MW: 153.58 g/mol
MDL Number: MFCD11506659
CAS Number: 1070896-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR