3,3-Dimethylpiperidin-4-ol

Supplier: Matrix Scientific

113236-1G 113236-5G
102891-718EA 213.85 USD
102891-718 102891-720
3,3-Dimethylpiperidin-4-ol
3,3-Dimethylpiperidin-4-ol

Hazard: Irritant

Formula: C7H15NO
MW: 129.2 g/mol
MDL Number: MFCD11044222
CAS Number: 373603-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR