3,3-Dimethylpiperidin-4-one hydrochloride

Supplier: Matrix Scientific

113235-1G 113235-5G
102891-714EA 727.69 USD
102891-714 102891-716
3,3-Dimethylpiperidin-4-one hydrochloride
3,3-Dimethylpiperidin-4-one hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO
MDL Number: MFCD11043097
CAS Number: 648921-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR