3-Fluoro-2-trifluoromethylisonicotinic acid

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-2-(trifluoromethyl)isonicotinic acid

113220-1G 113220-5G
102891-684EA 727.69 USD
102891-684 102891-686
3-Fluoro-2-trifluoromethylisonicotinic acid
3-Fluoro-2-trifluoromethylisonicotinic acid

Hazard: Irritant

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
MDL Number: MFCD08437597
CAS Number: 886510-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR