3-(Benzyloxy)cyclobutanol

Supplier: Matrix Scientific

113211-1G 113211-5G
102891-670EA 436.92 USD
102891-670 102891-672
3-(Benzyloxy)cyclobutanol
3-(Benzyloxy)cyclobutanol

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD21756137
CAS Number: 100058-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR