trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride

Supplier: Matrix Scientific

Synonyms: trans-4-Fluoropyrrolidin-3-ol hydrochloride

113196-1G 113196-5G
102891-636EA 960 USD
102891-636 102891-638
trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride
trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride

Hazard: Irritant

Formula: C₄H₈FNO·HCl
MW: 141.572 g/mol
MDL Number: MFCD23105899
CAS Number: 2006333-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR