cis-3,4-Dihydroxypyrrolidine hydrochloride

Supplier: Matrix Scientific

Synonyms: cis-Pyrrolidine-3,4-diol hydrochloride

113194-1G 113194-5G
102891-630EA 727.69 USD
102891-630 102891-632
cis-3,4-Dihydroxypyrrolidine hydrochloride
cis-3,4-Dihydroxypyrrolidine hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
MDL Number: MFCD21602660
CAS Number: 186393-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR