3-Trifluoromethyl-7-azaindole

Supplier: Matrix Scientific

113182-1G 113182-5G
102897-614EA 727.69 USD
102897-614 102897-616
3-Trifluoromethyl-7-azaindole
3-Trifluoromethyl-7-azaindole

Hazard: Irritant

Formula: C₈H₅F₃N₂
MDL Number: MFCD12024500
CAS Number: 892414-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR