7-Bromo-1,2,3,4-tetrahydroquinoline

Supplier: Matrix Scientific

113179-1G 113179-5G
102897-608EA 227.69 USD
102897-608 102897-610
7-Bromo-1,2,3,4-tetrahydroquinoline
7-Bromo-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₉H₁₀BrN
MW: 212.1 g/mol
MDL Number: MFCD08544270
CAS Number: 114744-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR