exo-8-Boc-8-azabicyclo[3.2.1]octane-3-carboxylic acid methyl ester

Supplier: Matrix Scientific

113164-1G 113164-5G
102897-768EA 727.69 USD
102897-768 102897-770
exo-8-Boc-8-azabicyclo[3.2.1]octane-3-carboxylic acid methyl ester
exo-8-Boc-8-azabicyclo[3.2.1]octane-3-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₄H₂₃NO₄
MDL Number: MFCD22415199
CAS Number: 1204809-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR