tert-Butyl-2-(oxetan-3-ylidene)acetate

Supplier: Matrix Scientific

113157-1G 113157-5G
102891-568EA 815.38 USD
102891-568 102891-612
tert-Butyl-2-(oxetan-3-ylidene)acetate
tert-Butyl-2-(oxetan-3-ylidene)acetate

Hazard: Irritant

Formula: C₉H₁₄O₃
MW: 170.21 g/mol
MDL Number: MFCD14586412
CAS Number: 1207175-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR