Ethyl-3-hydroxycyclobutanecarboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-hydroxycyclobutanecarboxylate

113147-1G 113147-5G
102891-554EA 190.77 USD
102891-554 102891-556
Ethyl-3-hydroxycyclobutanecarboxylate
Ethyl-3-hydroxycyclobutanecarboxylate

Hazard: Irritant

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD09758972
CAS Number: 17205-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR