6-Aminoisoindolin-1-one

Supplier: Matrix Scientific

Synonyms: 6-Amino-2,3-dihydro-1H-isoindol-1-one

113139-1G 113139-5G
102891-534EA 307.69 USD
102891-534 102891-536
6-Aminoisoindolin-1-one
6-Aminoisoindolin-1-one

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD09701282
CAS Number: 675109-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR