5-Bromo-3-chloro-2-methylpyridine

Supplier: Matrix Scientific

113126-1G 113126-5G
102891-510EA 838.46 USD
102891-510 102891-512
5-Bromo-3-chloro-2-methylpyridine
5-Bromo-3-chloro-2-methylpyridine

Hazard: Irritant

Formula: C₆H₅BrClN
MDL Number: MFCD09031417
CAS Number: 914358-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR