(3aR,6aR)-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-one

Supplier: Matrix Scientific

Synonyms: (3aR,6aR)-2,2-Dimethyl-3aH-cyclopenta[d][1,3]dioxol-4(6aH)-one

113124-5G 113124-1G
102891-506EA 1467.69 USD
102891-506 102897-340
(3aR,6aR)-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-one
(3aR,6aR)-3a,6a-Dihydro-2,2-dimethyl-4H-cyclopenta-1,3-dioxol-4-one

Hazard: Irritant

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
MDL Number: MFCD08166479
CAS Number: 115509-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR