2-Amino-4-iodopyridine

Supplier: Matrix Scientific

Synonyms: 4-Iodopyridin-2-amine

113121-1G 113121-5G
102897-332EA 436.92 USD
102897-332 102897-334
2-Amino-4-iodopyridine
2-Amino-4-iodopyridine

Hazard: Irritant

Formula: C₅H₅IN₂
MW: 220.01 g/mol
MDL Number: MFCD09038163
CAS Number: 552331-00-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR