1-Boc-3-(aminomethyl)-3-hydroxypyrrolidine
Supplier: Matrix Scientific
113113-1G
113113-5G
102897-712EA
664.62
USD
102897-712
102897-714
1-Boc-3-(aminomethyl)-3-hydroxypyrrolidine
1-Boc-3-(aminomethyl)-3-hydroxypyrrolidine
Hazard: Irritant
Formula:
C₁₀H₂₀N₂O₃ |
MDL Number:
MFCD18917057 CAS Number: 114214-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...