Risperidone

Supplier: Matrix Scientific

Synonyms: 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-(2-(4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one

113099-1G 113099-5G
102891-478EA 110.77 USD
102891-478 102891-480
Risperidone
Risperidone

Hazard: Irritant

Formula: C₂₃H₂₇FN₄O₂
MW: 410.49 g/mol
Melting Pt: 172…175 °C
MDL Number: MFCD00274576
CAS Number: 106266-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR