4-Chloro-2-methyl-6-(methylthio)pyrimidine

Supplier: Matrix Scientific

113098-1G 113098-5G
102891-474EA 561.54 USD
102891-474 102891-476
4-Chloro-2-methyl-6-(methylthio)pyrimidine
4-Chloro-2-methyl-6-(methylthio)pyrimidine

Hazard: Irritant

Formula: C₆H₇ClN₂S
MDL Number: MFCD16769133
CAS Number: 867131-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR