2,5-Dihydroxy-1,4-dioxane

Supplier: Matrix Scientific

Synonyms: 1,4-Dioxane-2,5-diol

113093-1G 113093-5G
102891-460EA 87.69 USD
102891-460 102891-462
2,5-Dihydroxy-1,4-dioxane
2,5-Dihydroxy-1,4-dioxane

Hazard: Irritant

Formula: C₄H₈O₄
MW: 120.11 g/mol
MDL Number: MFCD00012133
CAS Number: 23147-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR