Methyl-4-methyltetrahydro-2H-pyran-4-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 4-methyltetrahydro-2H-pyran-4-carboxylate

113084-1G 113084-5G
102897-702EA 189.23 USD
102897-702 102897-704
Methyl-4-methyltetrahydro-2H-pyran-4-carboxylate
Methyl-4-methyltetrahydro-2H-pyran-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD12912101
CAS Number: 443912-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR