Ethyl-1-boc-3-pyrrolidine acetate

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(2-ethoxy-2-oxoethyl)pyrrolidine-1-carboxylate

113081-1G 113081-5G
102897-694EA 461.54 USD
102897-694 102897-696
Ethyl-1-boc-3-pyrrolidine acetate
Ethyl-1-boc-3-pyrrolidine acetate

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
MDL Number: MFCD11977331
CAS Number: 664364-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR