(S)-1-Boc-3-bromomethylpiperidine

Supplier: Matrix Scientific

Synonyms: (S)-tert-Butyl 3-(bromomethyl)piperidine-1-carboxylate

113077-1G 113077-5G
102897-686EA 727.69 USD
102897-686 102897-688
(S)-1-Boc-3-bromomethylpiperidine
(S)-1-Boc-3-bromomethylpiperidine

Hazard: Irritant

Formula: C₁₁H₂₀BrNO₂
MW: 278.2 g/mol
MDL Number: MFCD11111933
CAS Number: 158406-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR