4-Bromo-1-(phenylsulfonyl)-1H-indazole

Supplier: Matrix Scientific

113069-1G 113069-5G
102897-676EA 256.92 USD
102897-676 102897-678
4-Bromo-1-(phenylsulfonyl)-1H-indazole
4-Bromo-1-(phenylsulfonyl)-1H-indazole

Hazard: Irritant

Formula: C₁₃H₉BrN₂O₂S
MDL Number: MFCD22566216
CAS Number: 1001415-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR