4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Supplier: Matrix Scientific

Synonyms: 4-bromo-1-(oxan-2-yl)-1H-indazole

113068-1G 113068-5G
102897-672EA 436.92 USD
102897-672 102897-674
4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Hazard: Irritant

Formula: C₁₂H₁₃BrN₂O
MW: 281.16 g/mol
MDL Number: MFCD22380249
CAS Number: 1022158-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR