4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine

Supplier: Matrix Scientific

113059-1G 113059-5G
102897-638EA 436.92 USD
102897-638 102897-640
4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine
4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₇H₆ClN₃
MDL Number: MFCD12924466
CAS Number: 871024-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR