(3-Ethyloxetan-3-yl)methanamine

Supplier: Matrix Scientific

113044-1G 113044-5G
102891-430EA 349.23 USD
102891-430 102891-432
(3-Ethyloxetan-3-yl)methanamine
(3-Ethyloxetan-3-yl)methanamine

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD11045879
CAS Number: 929900-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR