2-(1H-1,2,3-Triazol-1-yl)benzoic acid

Supplier: Matrix Scientific

113037-1G 113037-5G
102891-414EA 727.69 USD
102891-414 102891-416
2-(1H-1,2,3-Triazol-1-yl)benzoic acid
2-(1H-1,2,3-Triazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₉H₇N₃O₂
MDL Number: MFCD20486487
CAS Number: 1085458-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR