1-Boc-2-azetidinemethanol

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-(hydroxymethyl)azetidine-1-carboxylate

113032-1G 113032-5G
102891-404EA 290.77 USD
102891-404 102891-406
1-Boc-2-azetidinemethanol
1-Boc-2-azetidinemethanol

Hazard: Irritant

Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD03790936
CAS Number: 174346-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR